Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 7-Amino-4-methylcoumarin, 98%

Catalog No. AC218062500
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC218062500 250 mg Glass bottle
AC218060010 1 g Glass bottle
AC218060050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC218062500 Supplier Thermo Scientific Chemicals Supplier No. 218062500
  • $131.00
    $106.65 / Each of 1

    Save $24.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 26093-31-2 | C10H9NO2 | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26093-31-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00006868
InChI Key GLNDAGDHSLMOKX-UHFFFAOYSA-N
Synonym 7-amino-4-methylcoumarin, coumarin 120, 7-amino-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-amino-4-methyl, coumarin, 7-amino-4-methyl, 4-methyl-7-aminocoumarin, 7-amino-4-methyl-chromen-2-one, unii-ocy3jct44x, ccris 4961, ocy3jct44x
PubChem CID 92249
ChEBI CHEBI:51771
IUPAC Name 7-amino-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=CC(N)=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 223.0°C to 226.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: slightly soluble.
Formula Weight 175.19
Percent Purity 98%
Grade Extra Pure
Physical Form Crystalline Powder
Chemical Name or Material 7-Amino-4-methylcoumarin
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 247-454-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.