Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-amino-4-methylcoumarin-arachidonamide | 862913-13-1 | 99.5% | 10 MG
SDP

Catalog No. 5000233988
Encompass_Preferred

AMC Arachidonoyl Amide (AMC-AA) is an AMC-tagged fatty acid amide used as a fluorogenic substrate to measure fatty acid amide hydrolase (FAAH) activity in biochemical assays. It is supplied as a light yellow solid with high reported purity and requires low-temperature storage and dry-ice shipment.

  • High purity: 99.5%.
  • Molecular weight: 461.64 g/mol.
  • Chemical formula: C30H39NO3.
  • Suitable as a substrate for FAAH activity assays.
  • Stored at -80°C and shipped on dry ice.

Catalog No. 50-002-33988 Supplier Medchemexpress LLC Supplier No. HY12095710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AMC Arachidonoyl Amide (AMC-AA) is an AMC-tagged fatty acid amide used as a fluorogenic substrate to measure fatty acid amide hydrolase (FAAH) activity in biochemical assays. It is supplied as a light yellow solid with high reported purity and requires low-temperature storage and dry-ice shipment.

  • High purity: 99.5%.
  • Molecular weight: 461.64 g/mol.
  • Chemical formula: C30H39NO3.
  • Suitable as a substrate for FAAH activity assays.
  • Stored at -80°C and shipped on dry ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.