Learn More
Medchemexpress LLC HY-D0981 100mg Medchemexpress, 7-Amino-4-(trifluoromethyl)coumarin CAS:53518-15-3 Purity:>98%
SDP
Catalog No. 501933224
Encompass_Preferred
Medchemexpress, HY-D0981 100mg 7-Amino-4-(trifluoromethyl)coumarin CAS:53518-15-3 7-Amino-4-(trifluoromethyl)coumarin (Coumarin 151) is a fluorescent marker for the sensitive detection of proteinases. The excitation and emission wavelengths are 400 and 490 nm, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3224 Supplier Medchemexpress LLC Supplier No. HYD0981100MG
Medchemexpress, HY-D0981 100mg 7-Amino-4-(trifluoromethyl)coumarin CAS:53518-15-3 7-Amino-4-(trifluoromethyl)coumarin (Coumarin 151) is a fluorescent marker for the sensitive detection of proteinases. The excitation and emission wavelengths are 400 and 490 nm, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.