Learn More
Apexbio Technology LLC 7-Amino-4-(trifluoromethyl)coumarin 53518-15-3 500mg
SDP
Catalog No. 5000097332
Encompass_Preferred
7-Amino-4-trifluoromethyl coumarin (CAS 53518-15-3) is a small-molecule antagonist targeting serotonin receptor subtype 1E (HTR1E) galanin receptor 2 (GALR2) and gamma receptor subtype 2 (GalR2) It is designed to antagonize these receptors thereby modulating receptor-mediated signaling pathways 7-Amino-4-trifluoromethyl coumarin exerts its biological activity primarily through receptor antagonism and inhibition of hydroxysteroid 17-beta dehydrogenase 4 (HSD17B4) enzymatic activity In biochemical assays it demonstrates inhibitory activity toward HSD17B4 with reported bioactivity at approximately 21 M Based on these pharmacological properties 7-Amino-4-trifluoromethyl coumarin holds research potential in the investigation of protease activity receptor-mediated signaling and metabolic enzyme regulation
Catalog No. 50-000-97332 Supplier Apexbio Technology LLC Supplier No. A4714500
7-Amino-4-trifluoromethyl coumarin (CAS 53518-15-3) is a small-molecule antagonist targeting serotonin receptor subtype 1E (HTR1E) galanin receptor 2 (GALR2) and gamma receptor subtype 2 (GalR2) It is designed to antagonize these receptors thereby modulating receptor-mediated signaling pathways 7-Amino-4-trifluoromethyl coumarin exerts its biological activity primarily through receptor antagonism and inhibition of hydroxysteroid 17-beta dehydrogenase 4 (HSD17B4) enzymatic activity In biochemical assays it demonstrates inhibitory activity toward HSD17B4 with reported bioactivity at approximately 21 M Based on these pharmacological properties 7-Amino-4-trifluoromethyl coumarin holds research potential in the investigation of protease activity receptor-mediated signaling and metabolic enzyme regulation