Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 7-amino-5-bromoindole

Catalog No. CDS015232 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS015232 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS015232 Supplier Sigma Aldrich Supplier No. CDS015232100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl 3-methyl-2-butenoate is an α,α-unsaturated ester
  • It is reported as overripe fruity, ethereal odorant, responsible for the aroma of snake fruit
  • Rate coefficients for the reaction between the no3 radical and methyl 3-methyl-2-butenoate has been evaluated

Specifications

Molecular Formula C8H7BrN2
Linear Formula C8H7BrN2
Quantity 25 g
MDL Number MFCD02093961
Molecular Weight (g/mol) 211.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.