Learn More
AdipoGen 7-Amino-NBD
SDP
Catalog No. 502561817
Encompass_Preferred
Chemical. CAS 10199-91-4. Formula C6H4N4O3. MW 180.12. Synthetic. Fluorogenic amine. Building block and reagent for synthesis of fluorescent probes. Used for the preparation of fluorogenic substrates, chemosensors and probes, in which NBD-amine is the high fluoresence component after release. Shows constant fluorescence intensity from pH2-9. Spectral data Ex=472nm, Em=555nm TRIS buffer, pH7.8
Catalog No. 50-256-1817 Supplier AdipoGen Supplier No. CDXA0176M500
Chemical. CAS 10199-91-4. Formula C6H4N4O3. MW 180.12. Synthetic. Fluorogenic amine. Building block and reagent for synthesis of fluorescent probes. Used for the preparation of fluorogenic substrates, chemosensors and probes, in which NBD-amine is the high fluoresence component after release. Shows constant fluorescence intensity from pH2-9. Spectral data Ex=472nm, Em=555nm TRIS buffer, pH7.8