Learn More
Medchemexpress LLC 7-Aminoactinomycin D | 7240-37-1 | ≥99.0% | 1270.43 | 1 MG
SDP
Catalog No. 5000429612
Encompass_Preferred
7-Aminoactinomycin D (7-AAD) is a cell-impermeant fluorescent DNA stain and a potent RNA polymerase inhibitor. It selectively binds to GC regions of DNA and also exhibits antibacterial effects. It is used as a DNA dye to distinguish viable, apoptotic, and late apoptotic/dead cells in flow cytometry.
  • Cell-impermeant fluorescent DNA stain.
  • Potent RNA polymerase inhibitor.
  • Selectively binds to GC regions of DNA.
  • Exhibits antibacterial effects.
  • Distinguishes viable, apoptotic, and late apoptotic/dead cells in flow cytometry.
  • Can be used at different concentrations for specific staining purposes in flow cytometry.
Catalog No. 50-004-29612 Supplier Medchemexpress LLC Supplier No. HYD10201MG
7-Aminoactinomycin D (7-AAD) is a cell-impermeant fluorescent DNA stain and a potent RNA polymerase inhibitor. It selectively binds to GC regions of DNA and also exhibits antibacterial effects. It is used as a DNA dye to distinguish viable, apoptotic, and late apoptotic/dead cells in flow cytometry.
  • Cell-impermeant fluorescent DNA stain.
  • Potent RNA polymerase inhibitor.
  • Selectively binds to GC regions of DNA.
  • Exhibits antibacterial effects.
  • Distinguishes viable, apoptotic, and late apoptotic/dead cells in flow cytometry.
  • Can be used at different concentrations for specific staining purposes in flow cytometry.