Learn More
Medchemexpress LLC HY-W014217 500mg Medchemexpress, 7-Aminodeacetoxycephalosporanic acid CAS:22252-43-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW014217500MG
Encompass_Preferred
Medchemexpress, HY-W014217 500mg 7-Aminodeacetoxycephalosporanic acid CAS:22252-43-3 7-Aminodeacetoxycephalosporanic acid (7-ADCA) is a key intermediate in the synthesis of cephalosporins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3358