Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Aminoheptanoic acid | 929-17-9 | MFCD00008242 | 145.20 | 10 G
SDP

Catalog No. 5000446485
Encompass_Preferred

7-Aminoheptanoic acid is a PROTAC linker used in the synthesis of PROTAC molecules. This product is intended for research use only.

  • White to off-white solid appearance
  • Molecular weight of 145.20
  • Purity of 98.0%
  • Stable at room temperature for 3 years
  • Stable in solvent at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-46485 Supplier Medchemexpress LLC Supplier No. HYW00204210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Aminoheptanoic acid is a PROTAC linker used in the synthesis of PROTAC molecules. This product is intended for research use only.

  • White to off-white solid appearance
  • Molecular weight of 145.20
  • Purity of 98.0%
  • Stable at room temperature for 3 years
  • Stable in solvent at -80°C for 2 years or -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.