Learn More
Medchemexpress LLC (7-aminoheptyl)-guanidine, monosulfate | 150417-90-6 | 98.0% | 270.35 | C8H22N4O4S | 100 MG
SDP
Catalog No. 5000244004
Encompass_Preferred
GC7 Sulfate is the sulfate salt of N1-guanyl-1,7-diaminoheptane, a selective deoxyhypusine synthase (DHPS) inhibitor used in vitro to study eIF5A hypusination, cell cycle regulation, and polyamine biology. It is supplied as a high-purity solid for research use only.
  • Selective deoxyhypusine synthase inhibitor for mechanistic studies.
  • Useful for probing eIF5A hypusination and polyamine-related pathways.
  • Demonstrated dose-dependent reduction of cell viability in multiple cell lines.
  • High purity (≥98.0%) solid, soluble in water for solution preparation.
  • Recommended storage: 4°C for solid; in solution -80°C (up to 6 months).
Catalog No. 50-002-44004 Supplier Medchemexpress LLC Supplier No. HY108314A100MG
GC7 Sulfate is the sulfate salt of N1-guanyl-1,7-diaminoheptane, a selective deoxyhypusine synthase (DHPS) inhibitor used in vitro to study eIF5A hypusination, cell cycle regulation, and polyamine biology. It is supplied as a high-purity solid for research use only.
  • Selective deoxyhypusine synthase inhibitor for mechanistic studies.
  • Useful for probing eIF5A hypusination and polyamine-related pathways.
  • Demonstrated dose-dependent reduction of cell viability in multiple cell lines.
  • High purity (≥98.0%) solid, soluble in water for solution preparation.
  • Recommended storage: 4°C for solid; in solution -80°C (up to 6 months).