Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Aminoquinoline, 97%

Catalog No. AC460050010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460050010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460050010 Supplier Thermo Scientific Chemicals Supplier No. 460050010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 580-19-8 | C9H8N2 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-19-8
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.17
InChI Key RZAUIOKDXQWSQE-UHFFFAOYSA-N
Synonym 7-aminoquinoline, 7-quinolinamine, 7-quinolylamine, quinoline, 7-amino, 7-amino-quinoline, ccris 1682, 7-quinolinamine 9ci, 7-aminequinolin, 7-aminequinoline, 7-quinolinamine acetate
PubChem CID 11377
IUPAC Name quinolin-7-amine
SMILES C1=CC2=C(C=C(C=C2)N)N=C1

Specifications

Melting Point 80.0°C to 86.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 144.17
Percent Purity 97%
Chemical Name or Material 7-Aminoquinoline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.