Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Anilino-4-hydroxy-2-naphthalenesulfonic Acid 95.0+%, TCI America™
SDP

Catalog No. A046725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A046725G 25 g
A0467500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A046725G Supplier TCI America Supplier No. A046725G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 119-40-4
Molecular Formula C16H13NO4S
Molecular Weight (g/mol) 315.343
MDL Number MFCD00035714
InChI Key YGNDWDUEMICDLW-UHFFFAOYSA-N
Synonym 7-Anilino-4-naphthol-2-sulfonic Acid, Phenyl J Acid
PubChem CID 67065
IUPAC Name 7-anilino-4-hydroxynaphthalene-2-sulfonic acid
SMILES C1=CC=C(C=C1)NC2=CC3=CC(=CC(=C3C=C2)O)S(=O)(=O)O

Specifications

Quantity 25 g
Formula Weight 315.34
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Anilino-4-hydroxy-2-naphthalenesulfonic Acid
EINECSNumber (4)-0536
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.