Learn More
Cayman Chemical 7-APBhydrochlorIde 10mg
SDP
Catalog No. 502953962
Encompass_Preferred
An analytical reference standard categorized as a benzofuran; a potential metabolite of 7-MAPB based on the published metabolism of 5-MAPB; intended for research and forensic applications
Catalog No. 50-295-3962 Supplier Cayman Chemical Supplier No. 1428210 MG
An analytical reference standard categorized as a benzofuran; a potential metabolite of 7-MAPB based on the published metabolism of 5-MAPB; intended for research and forensic applications