Learn More
Medchemexpress LLC 7-APRA | 120709-09-3 | ≥95.0% | 240.28 | 10 G
SDP
Catalog No. 5000348172
Encompass_Preferred
7-APRA is a semi-synthetic intermediate of cephalosporin antibiotics. It possesses antibacterial activity and is primarily utilized in the synthesis of Cefaclor and Cefprozil.
  • Possesses antibacterial activity
  • Used in the synthesis of cephalosporin antibiotics
  • Intermediate for drug synthesis
Catalog No. 50-003-48172 Supplier Medchemexpress LLC Supplier No. HY13817910G
7-APRA is a semi-synthetic intermediate of cephalosporin antibiotics. It possesses antibacterial activity and is primarily utilized in the synthesis of Cefaclor and Cefprozil.
  • Possesses antibacterial activity
  • Used in the synthesis of cephalosporin antibiotics
  • Intermediate for drug synthesis