Learn More
Medchemexpress LLC 7-APRA | 120709-09-3 | ≥95.0% | 240.28 | 25 G
SDP
Catalog No. 5000351572
Encompass_Preferred
7-APRA is a semi-synthetic intermediate of cephalosporin antibiotics. It possesses antibacterial activity and is primarily utilized in the synthesis of Cefaclor and Cefprozil. It appears as an off-white to light yellow solid.
  • Possesses antibacterial activity
  • Utilized in the synthesis of Cefaclor and Cefprozil
  • Solid appearance
  • Off-white to light yellow color
Catalog No. 50-003-51572 Supplier Medchemexpress LLC Supplier No. 25GHY138179
Add to Cart
Add to Cart
7-APRA is a semi-synthetic intermediate of cephalosporin antibiotics. It possesses antibacterial activity and is primarily utilized in the synthesis of Cefaclor and Cefprozil. It appears as an off-white to light yellow solid.
  • Possesses antibacterial activity
  • Utilized in the synthesis of Cefaclor and Cefprozil
  • Solid appearance
  • Off-white to light yellow color