Learn More
Medchemexpress LLC 7-APRA | 120709-09-3 | ≥95.0% | 240.28 | 50 G
SDP
Catalog No. 5000347777
Encompass_Preferred
7-APRA is a semi-synthetic intermediate of cephalosporin antibiotics. It possesses antibacterial activity and is primarily utilized in the synthesis of Cefaclor and Cefprozil.
  • Semi-synthetic intermediate of cephalosporin antibiotics
  • Has antibacterial activity
  • Used in synthesis of other compounds
  • Appears as a solid
  • Color is off-white to light yellow
Catalog No. 50-003-47777 Supplier Medchemexpress LLC Supplier No. HY13817950G
Add to Cart
Add to Cart
7-APRA is a semi-synthetic intermediate of cephalosporin antibiotics. It possesses antibacterial activity and is primarily utilized in the synthesis of Cefaclor and Cefprozil.
  • Semi-synthetic intermediate of cephalosporin antibiotics
  • Has antibacterial activity
  • Used in synthesis of other compounds
  • Appears as a solid
  • Color is off-white to light yellow