Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Azaindole | 271-63-6 | 99.99% | C7H6N2 | 25 G
SDP

Catalog No. 5000359041
Encompass_Preferred

7-Azaindole, also known as 1H-Pyrrolo[2,3-b]pyridine, is a biochemical reagent primarily used in life science related research. This compound serves as a biological material or organic compound for various laboratory applications. It is supplied as a high-purity, white to off-white solid.

  • High purity: 99.99%
  • Appearance: White to off-white solid
  • Molecular weight: 118.14
  • Chemical formula: C7H6N2
  • Suitable for life science research

Catalog No. 50-003-59041 Supplier Medchemexpress LLC Supplier No. 25GHY50542
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Azaindole, also known as 1H-Pyrrolo[2,3-b]pyridine, is a biochemical reagent primarily used in life science related research. This compound serves as a biological material or organic compound for various laboratory applications. It is supplied as a high-purity, white to off-white solid.

  • High purity: 99.99%
  • Appearance: White to off-white solid
  • Molecular weight: 118.14
  • Chemical formula: C7H6N2
  • Suitable for life science research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.