Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 7-Azaindole-5-boronic acid pinacol ester, >95%, [754214-56-7], A13809 244.097 C13H17BN2O2

Catalog No. 501374548
Encompass_Preferred

Compound: 7-Azaindole-5-boronic acid pinacol ester

CAS#: 754214-56-7

MDL#: not listed

Background: An azaindole boronic ester used as a Suzuki-Miyaura coupling partner in pharmaceutical research. Azaindoles are privileged nitrogen heterocyclic scaffolds in drug discovery, and this ester provides direct access to substituted azaindole building blocks.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-137-4548 Supplier Frontier Specialty Chemicals Supplier No. A138095G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 7-Azaindole-5-boronic acid pinacol ester

CAS#: 754214-56-7

MDL#: not listed

Background: An azaindole boronic ester used as a Suzuki-Miyaura coupling partner in pharmaceutical research. Azaindoles are privileged nitrogen heterocyclic scaffolds in drug discovery, and this ester provides direct access to substituted azaindole building blocks.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.