Learn More
Medchemexpress LLC 7-azido-4-methyl-2H-1-benzopyran-2-one | 95633-27-5 | 98.9% | 100 MG
SDP
Catalog No. 5000233650
Encompass_Preferred
7-Azido-4-methylcoumarin is a coumarin-derived fluorogenic probe that becomes fluorescent upon reduction of the azido group, enabling sensitive detection of hydrogen sulfide (H2S) in biochemical assays. It is supplied as an off-white to light-yellow solid for research use.
  • Fluorogenic H2S probe that yields a fluorescent amine upon azide reduction.
  • High purity suitable for biochemical assays.
  • Off-white to light-yellow solid form for convenient handling.
  • Stable as a powder at -20°C for extended storage.
  • Available in small research-scale quantities (100 MG).
  • Molecular weight 201.18 g/mol; formula C10H7N3O2.
Catalog No. 50-002-33650 Supplier Medchemexpress LLC Supplier No. HY119323100MG
7-Azido-4-methylcoumarin is a coumarin-derived fluorogenic probe that becomes fluorescent upon reduction of the azido group, enabling sensitive detection of hydrogen sulfide (H2S) in biochemical assays. It is supplied as an off-white to light-yellow solid for research use.
  • Fluorogenic H2S probe that yields a fluorescent amine upon azide reduction.
  • High purity suitable for biochemical assays.
  • Off-white to light-yellow solid form for convenient handling.
  • Stable as a powder at -20°C for extended storage.
  • Available in small research-scale quantities (100 MG).
  • Molecular weight 201.18 g/mol; formula C10H7N3O2.