Learn More
Cayman Chemical 7-Azido-4-methylcoumarin, Purity: ≥95%, CAS: 95633-27-5, 5 mg
SDP
Catalog No. 502939074
Encompass_Preferred
A fluorescent H2S probe; ex/em = 340/445 nm, respectively; selectively fluoresces in the presence of H2S over a panel of reactive nitrogen, oxygen, and sulfur species
Catalog No. 50-293-9074 Supplier Cayman Chemical Supplier No. 283075 MG
A fluorescent H2S probe; ex/em = 340/445 nm, respectively; selectively fluoresces in the presence of H2S over a panel of reactive nitrogen, oxygen, and sulfur species