Learn More
Medchemexpress LLC 7-azido-4-methylcoumarin | 95633-27-5 | 98.9% | 5 MG
SDP
Catalog No. 5000234675
Encompass_Preferred
7-Azido-4-methylcoumarin is a coumarin-based fluorogenic probe used to detect hydrogen sulfide (H2S) and to monitor cystathionine β-synthase activity. The molecule contains an azide functional group that is reducible by H2S and can participate in bioorthogonal click chemistry.
  • Fluorogenic probe for hydrogen sulfide (H2S) detection.
  • Useful for assays of cystathionine β-synthase activity.
  • Contains an azide functional group reactive in CuAAC and SPAAC click chemistry.
  • Available as solid packs (including 5 mg) and as a 10 mM solution in DMSO.
  • Offered with high reported purity (98.9%) and molecular weight 201.18 g/mol.
Catalog No. 50-002-34675 Supplier Medchemexpress LLC Supplier No. HY1193235MG
7-Azido-4-methylcoumarin is a coumarin-based fluorogenic probe used to detect hydrogen sulfide (H2S) and to monitor cystathionine β-synthase activity. The molecule contains an azide functional group that is reducible by H2S and can participate in bioorthogonal click chemistry.
  • Fluorogenic probe for hydrogen sulfide (H2S) detection.
  • Useful for assays of cystathionine β-synthase activity.
  • Contains an azide functional group reactive in CuAAC and SPAAC click chemistry.
  • Available as solid packs (including 5 mg) and as a 10 mM solution in DMSO.
  • Offered with high reported purity (98.9%) and molecular weight 201.18 g/mol.