Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Benzyl-2,7-diazaspiro[3.5]nonane dihydrochloride | 1334536-89-8 | 95% | 289.24 | 5 G
SDP

Catalog No. 5000401221
Encompass_Preferred

7-Benzyl-2,7-diazaspiro[3.5]nonane dihydrochloride is a drug intermediate for synthesis of various active compounds.

  • Molecular formula: C14H22Cl2N2
  • Target: Drug intermediate
  • Pathway: Others
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Application: Reference compound

Catalog No. 50-004-01221 Supplier Medchemexpress LLC Supplier No. HY30761A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Benzyl-2,7-diazaspiro[3.5]nonane dihydrochloride is a drug intermediate for synthesis of various active compounds.

  • Molecular formula: C14H22Cl2N2
  • Target: Drug intermediate
  • Pathway: Others
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Application: Reference compound

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.