Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-(Benzyloxy)-3,4-dihydronaphthalen-1(2H)-one | 32263-64-2 | 99.7% | C17H16O2 | 500 MG
SDP

Catalog No. 5000357761
Encompass_Preferred

7-(Benzyloxy)-3,4-dihydronaphthalen-1(2H)-one is a drug intermediate for the synthesis of various active compounds. This product is intended for research use only and is not sold to patients.

  • A drug intermediate for the synthesis of various active compounds.
  • Intended for research use only.
  • Appearance: Solid.
  • Molecular weight: 252.31.
  • Shipping: Room temperature in continental US.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-003-57761 Supplier Medchemexpress LLC Supplier No. HY47241500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-(Benzyloxy)-3,4-dihydronaphthalen-1(2H)-one is a drug intermediate for the synthesis of various active compounds. This product is intended for research use only and is not sold to patients.

  • A drug intermediate for the synthesis of various active compounds.
  • Intended for research use only.
  • Appearance: Solid.
  • Molecular weight: 252.31.
  • Shipping: Room temperature in continental US.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.