Learn More
Medchemexpress LLC 7-(Benzyloxy)-6-methoxyquinazolin-4(3H)-one | 179688-01-8 | MFCD04115119 | >98.0% | 282.30 | 10 G
SDP
Catalog No. 5000395962
Encompass_Preferred
7-(Benzyloxy)-6-methoxyquinazolin-4(3H)-one is a biochemical reagent presented as a white to almost white powder or crystal. This compound has a purity of >98.0% and a molecular weight of 282.30. It is suitable for various laboratory applications.
  • Solid physical state at 20 °C
  • White to almost white powder or crystal appearance
  • Melting point of 269 °C
Catalog No. 50-003-95962 Supplier Medchemexpress LLC Supplier No. HYW00853110G
7-(Benzyloxy)-6-methoxyquinazolin-4(3H)-one is a biochemical reagent presented as a white to almost white powder or crystal. This compound has a purity of >98.0% and a molecular weight of 282.30. It is suitable for various laboratory applications.
  • Solid physical state at 20 °C
  • White to almost white powder or crystal appearance
  • Melting point of 269 °C