Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-BIA 100mg | 1313403-49-4 | 294.30 g·mol⁻1 | C15H18O6 | 100 MG
SDP

Catalog No. 5000229146
Encompass_Preferred

7-BIA is a small-molecule inhibitor of receptor-type protein tyrosine phosphatase D (PTPRD), reported to inhibit PTPRD with an IC50 of approximately 1-3 μM. It is supplied as a high-purity solid research reagent for biochemical and cellular studies.

  • Inhibits PTPRD (IC50 ≈ 1-3 μM) and PTPRS (IC50 40 μM).
  • High purity: >98.0% (HPLC).
  • Molecular formula C15H18O6; molecular weight 294.30 g·mol⁻1.
  • Physical form: solid, off-white to light yellow.
  • CAS number 1313403-49-4 for unambiguous identification.
  • Available as 100 MG solid and alternative pack sizes for research use.

Catalog No. 50-002-29146 Supplier Medchemexpress LLC Supplier No. HY115496100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-BIA is a small-molecule inhibitor of receptor-type protein tyrosine phosphatase D (PTPRD), reported to inhibit PTPRD with an IC50 of approximately 1-3 μM. It is supplied as a high-purity solid research reagent for biochemical and cellular studies.

  • Inhibits PTPRD (IC50 ≈ 1-3 μM) and PTPRS (IC50 40 μM).
  • High purity: >98.0% (HPLC).
  • Molecular formula C15H18O6; molecular weight 294.30 g·mol⁻1.
  • Physical form: solid, off-white to light yellow.
  • CAS number 1313403-49-4 for unambiguous identification.
  • Available as 100 MG solid and alternative pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.