Learn More
Medchemexpress LLC 7-BIA 10mg | 1313403-49-4 | 294.30 g/mol | C15H18O6 | 10 MG
SDP
Catalog No. 5000307992
Encompass_Preferred
7-BIA is a small-molecule inhibitor of receptor-type protein tyrosine phosphatase D (PTPRD) used for biochemical and cellular research. Reported activity: PTPRD IC50 ≈ 1-3 μM; PTPRS IC50 = 40 μM. Molecular formula C15H18O6; molecular weight 294.30 g/mol; purity 98.0%.
  • Potent PTPRD inhibition (IC50 ≈ 1-3 μM)
  • Also inhibits PTPRS (IC50 = 40 μM)
  • High stated purity of 98.0%
  • Supplied as solid and solution formats for flexible use
  • Suitable for biochemical and cellular research applications
Catalog No. 50-003-07992 Supplier Medchemexpress LLC Supplier No. HY11549610MG
7-BIA is a small-molecule inhibitor of receptor-type protein tyrosine phosphatase D (PTPRD) used for biochemical and cellular research. Reported activity: PTPRD IC50 ≈ 1-3 μM; PTPRS IC50 = 40 μM. Molecular formula C15H18O6; molecular weight 294.30 g/mol; purity 98.0%.
  • Potent PTPRD inhibition (IC50 ≈ 1-3 μM)
  • Also inhibits PTPRS (IC50 = 40 μM)
  • High stated purity of 98.0%
  • Supplied as solid and solution formats for flexible use
  • Suitable for biochemical and cellular research applications