Learn More
Medchemexpress LLC 7-BIA 50mg | 1313403-49-4 | 294.30 g/mol | C15H18O6 | 50 MG
SDP
Catalog No. 5000226730
Encompass_Preferred
7-BIA is a small-molecule inhibitor of receptor-type protein tyrosine phosphatase D (PTPRD) with reported IC50 ≈ 1-3 μM. Supplied for laboratory research, it is available as solids and as ready-to-use DMSO solutions for biochemical and cellular assays.
  • Potent inhibitor of PTPRD with IC50 ≈ 1-3 μM.
  • High purity (98.0%) suitable for biochemical assays.
  • Molecular formula C15H18O6 and molecular weight 294.30 g/mol.
  • Available as solid and as 10 mM solution in DMSO for convenient use.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-26730 Supplier Medchemexpress LLC Supplier No. HY11549650MG
7-BIA is a small-molecule inhibitor of receptor-type protein tyrosine phosphatase D (PTPRD) with reported IC50 ≈ 1-3 μM. Supplied for laboratory research, it is available as solids and as ready-to-use DMSO solutions for biochemical and cellular assays.
  • Potent inhibitor of PTPRD with IC50 ≈ 1-3 μM.
  • High purity (98.0%) suitable for biochemical assays.
  • Molecular formula C15H18O6 and molecular weight 294.30 g/mol.
  • Available as solid and as 10 mM solution in DMSO for convenient use.
  • Intended for research use only; not for human or clinical use.