Learn More
Cayman Chemical 7-BIopterIn 10mg
SDP
Catalog No. 502989713
Encompass_Preferred
A pterin; formed by non-enzymatic rearrangement of 4a-hydroxy-tetrahydropterin in the absence of PCD in vitro and levels are elevated in the urine of hyperphenylalaninemia patients carrying heterozygous mutations in the PCBD gene encoding PCD
Catalog No. 50-298-9713 Supplier Cayman Chemical Supplier No. 2981910 MG
A pterin; formed by non-enzymatic rearrangement of 4a-hydroxy-tetrahydropterin in the absence of PCD in vitro and levels are elevated in the urine of hyperphenylalaninemia patients carrying heterozygous mutations in the PCBD gene encoding PCD