Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 7-Bromo-2-methyl-2H-indazole

Catalog No. SYX001291G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX001291G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX001291G Supplier Sigma Aldrich Supplier No. SYX001291G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-methylcyclopentene can be used as:
  • A probe molecule in the synthesis of alkenyl carbenium ions via adsorption on zeolite y
  • A model olefin compound in the kinetic study of hydrogenation of olefins using various catalytic systems
  • A reactant to synthesize 1,2,3-trisubstituted
  • Benzocyclopentenes by reacting with α-disulfonyl iodonium ylides

Specifications

CAS 701910-14-7
Molecular Formula C8H7BrN2
Linear Formula C8H7BrN2
MDL Number MFCD09870049
Quantity 5 g
Molecular Weight (g/mol) 211.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.