Learn More
eMolecules​ 302912-38-5 | 7-Bromo-2-methylindole | Combi-Blocks, Inc. | MFCD02683530 | 210.074 | C9H8BrN | 96.000 | Cc1cc2cccc(Br)c2[nH]1 | 1g | 603182116
Catalog No. NC2226259 Shop All eMolecules Inc. Products
Encompass_Preferred
7-Bromo-2-methylindole | Combi-Blocks, Inc. | 302912-38-5 | MFCD02683530 | 210.074 | C9H8BrN | 96.000 | Cc1cc2cccc(Br)c2[nH]1 | 1g | 603182116
This item is not returnable. View return policy
Catalog No. NC2226259 Supplier eMolecules​ Supplier No. 603182116
7-Bromo-2-methylindole | Combi-Blocks, Inc. | 302912-38-5 | MFCD02683530 | 210.074 | C9H8BrN | 96.000 | Cc1cc2cccc(Br)c2[nH]1 | 1g | 603182116