Learn More
Medchemexpress LLC 7-Bromoheptanoic acid | 30515-28-7 | 209.08 | 1 ML
SDP
Catalog No. 5000446587
Encompass_Preferred
7-Bromoheptanoic acid is a biochemical reagent suitable for life science-related research as a biological material or organic compound.
  • It can be used in the synthesis of azide-based nicotinamide phosphoribosyltransferase inhibitors with anticancer activity.
  • It has been utilized in the synthesis of derivatives that inhibit histone deacetylases.
Catalog No. 50-004-46587 Supplier Medchemexpress LLC Supplier No. HYW00771410MM/1ML
7-Bromoheptanoic acid is a biochemical reagent suitable for life science-related research as a biological material or organic compound.
  • It can be used in the synthesis of azide-based nicotinamide phosphoribosyltransferase inhibitors with anticancer activity.
  • It has been utilized in the synthesis of derivatives that inhibit histone deacetylases.