Learn More
Medchemexpress LLC 7-Bromoheptanoic acid | 30515-28-7 | 209.08 | 10 G
SDP
Catalog No. 5000448699
Encompass_Preferred
7-Bromoheptanoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It acts as a versatile building block in various synthetic applications.
  • Utilized in the synthesis of azide-based nicotinamide phosphoribosyltransferase (Nampt) inhibitors with anticancer activity.
  • Used in the synthesis of SAHA derivatives that inhibit histone deacetylases (HDACs).
  • For research use only.
Catalog No. 50-004-48699 Supplier Medchemexpress LLC Supplier No. HYW00771410G
7-Bromoheptanoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It acts as a versatile building block in various synthetic applications.
  • Utilized in the synthesis of azide-based nicotinamide phosphoribosyltransferase (Nampt) inhibitors with anticancer activity.
  • Used in the synthesis of SAHA derivatives that inhibit histone deacetylases (HDACs).
  • For research use only.