Learn More
Medchemexpress LLC 7-bromoindirubin-3-oxime | 916440-85-2 | 99.2% | 5 MG
SDP
Catalog No. 5000232044
Encompass_Preferred
7-bromoindirubin-3-oxime is an indirubin-derived, kinase-inhibiting research compound used in preclinical studies to probe CDK5 and GSK3β signaling and Aβ-related neurotoxicity.
  • Inhibits CDK5 and GSK3β, useful for kinase pathway studies.
  • Reported to reduce Aβ oligomer-induced neuroinflammation and synaptic impairment in models.
  • High purity appropriate for biochemical and cellular assays.
  • Stable as a solid when stored at 4°C and protected from light.
  • Available in small milligram pack sizes for research use.
Catalog No. 50-002-32044 Supplier Medchemexpress LLC Supplier No. HY1210355MG
7-bromoindirubin-3-oxime is an indirubin-derived, kinase-inhibiting research compound used in preclinical studies to probe CDK5 and GSK3β signaling and Aβ-related neurotoxicity.
  • Inhibits CDK5 and GSK3β, useful for kinase pathway studies.
  • Reported to reduce Aβ oligomer-induced neuroinflammation and synaptic impairment in models.
  • High purity appropriate for biochemical and cellular assays.
  • Stable as a solid when stored at 4°C and protected from light.
  • Available in small milligram pack sizes for research use.