Learn More
Medchemexpress LLC 7-bromoquinoline-4-carboxylic acid | 31009-04-8 | MFCD11108751 | 96.1% | 252.06 | C10H6BrNO2 | 1 G
SDP
Catalog No. 5000300134
Encompass_Preferred
7-Bromoquinoline-4-carboxylic acid is a brominated quinoline carboxylic acid used as an intermediate in organic synthesis and medicinal chemistry research. It is supplied as a gray to brown solid (C10H6BrNO2; MW 252.06; CAS 31009-04-8) with typical HPLC purity of 96.12%. Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Used as an intermediate for organic synthesis
  • Suitable for medicinal chemistry research applications
  • Supplied as a stable solid with defined storage conditions
  • Provided with HPLC purity data for quality assessment
  • Batch certificate of analysis available for traceability
Catalog No. 50-003-00134 Supplier Medchemexpress LLC Supplier No. HYW0221641G
7-Bromoquinoline-4-carboxylic acid is a brominated quinoline carboxylic acid used as an intermediate in organic synthesis and medicinal chemistry research. It is supplied as a gray to brown solid (C10H6BrNO2; MW 252.06; CAS 31009-04-8) with typical HPLC purity of 96.12%. Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Used as an intermediate for organic synthesis
  • Suitable for medicinal chemistry research applications
  • Supplied as a stable solid with defined storage conditions
  • Provided with HPLC purity data for quality assessment
  • Batch certificate of analysis available for traceability