Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-bromoquinoline-4-carboxylic acid | 31009-04-8 | MFCD11108751 | 96.1% | 252.06 | C10H6BrNO2 | 50 MG
SDP

Catalog No. 5000300502
Encompass_Preferred

7-Bromoquinoline-4-carboxylic acid is a quinoline-derived carboxylic acid supplied as a solid powder and used primarily as an intermediate in organic synthesis and medicinal chemistry research. It is suitable for building small-molecule libraries, derivatization, and further functionalization in synthetic workflows.

  • Used as an intermediate in organic and medicinal chemistry synthesis.
  • Powder form suitable for cold storage and handling.
  • Characterized by molecular formula C10H6BrNO2 and molecular weight 252.06.
  • Purity 96.1% determined by HPLC.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term).

Catalog No. 50-003-00502 Supplier Medchemexpress LLC Supplier No. HYW02216450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Bromoquinoline-4-carboxylic acid is a quinoline-derived carboxylic acid supplied as a solid powder and used primarily as an intermediate in organic synthesis and medicinal chemistry research. It is suitable for building small-molecule libraries, derivatization, and further functionalization in synthetic workflows.

  • Used as an intermediate in organic and medicinal chemistry synthesis.
  • Powder form suitable for cold storage and handling.
  • Characterized by molecular formula C10H6BrNO2 and molecular weight 252.06.
  • Purity 96.1% determined by HPLC.
  • Storage recommendations: powder at -20°C (long term) or 4°C (short term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.