Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-bromoquinoline-4-carboxylic acid | 31009-04-8 | MFCD11108751 | 96.1% | 252.06 | C10H6BrNO2 | 500 MG
SDP

Catalog No. 5000300967
Encompass_Preferred

7-Bromoquinoline-4-carboxylic acid is an organic small-molecule reagent used primarily as an intermediate in synthetic chemistry and for biochemical assay research. It is a brominated quinoline carboxylic acid commonly employed to build substituted heterocycles and as a starting material for structure-activity studies.

  • Used as an intermediate in chemical synthesis and biochemical research.
  • Molecular formula C10H6BrNO2 and molecular weight 252.06 g·mol-1.
  • CAS number 31009-04-8 for unambiguous identification.
  • Typical purity 96.1% (HPLC) as reported on the certificate of analysis.
  • Appearance: gray to brown solid.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-00967 Supplier Medchemexpress LLC Supplier No. HYW022164500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Bromoquinoline-4-carboxylic acid is an organic small-molecule reagent used primarily as an intermediate in synthetic chemistry and for biochemical assay research. It is a brominated quinoline carboxylic acid commonly employed to build substituted heterocycles and as a starting material for structure-activity studies.

  • Used as an intermediate in chemical synthesis and biochemical research.
  • Molecular formula C10H6BrNO2 and molecular weight 252.06 g·mol-1.
  • CAS number 31009-04-8 for unambiguous identification.
  • Typical purity 96.1% (HPLC) as reported on the certificate of analysis.
  • Appearance: gray to brown solid.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.