Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-bromoquinoline-4-carboxylic acid | 31009-04-8 | MFCD11108751 | 96.1% | 252.06 g·mol⁻1 | C10H6BrNO2 | 100 MG
SDP

Catalog No. 5000299528
Encompass_Preferred

7-Bromoquinoline-4-carboxylic acid is a bromo-substituted quinoline carboxylic acid used as an intermediate and reagent in medicinal chemistry and organic synthesis. Manufacturer batch documentation provides molecular, analytical, and storage information to support research use and quality control.

  • Contains a bromine-substituted quinoline core suitable for derivatization.
  • Useful intermediate for medicinal chemistry and organic synthesis.
  • Molecular formula C10H6BrNO2 and molecular weight 252.06 g·mol⁻1.
  • Solid powder appearance; batch color may be gray to brown.
  • Analytical data available, including HPLC purity and NMR confirmation.
  • Recommended storage: powder at -20 °C for long term, 4 °C for shorter term; solutions at -80 °C for extended storage.

Catalog No. 50-002-99528 Supplier Medchemexpress LLC Supplier No. HYW022164100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Bromoquinoline-4-carboxylic acid is a bromo-substituted quinoline carboxylic acid used as an intermediate and reagent in medicinal chemistry and organic synthesis. Manufacturer batch documentation provides molecular, analytical, and storage information to support research use and quality control.

  • Contains a bromine-substituted quinoline core suitable for derivatization.
  • Useful intermediate for medicinal chemistry and organic synthesis.
  • Molecular formula C10H6BrNO2 and molecular weight 252.06 g·mol⁻1.
  • Solid powder appearance; batch color may be gray to brown.
  • Analytical data available, including HPLC purity and NMR confirmation.
  • Recommended storage: powder at -20 °C for long term, 4 °C for shorter term; solutions at -80 °C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.