Learn More
Medchemexpress LLC 7-chloro-3-(2,4-dimethoxyphenyl)-5-methyl-thiazolo[4,5-d]pyrimidine-2-thione | 2459916-56-2 | 98.5% | 353.85 g/mol | C14H12ClN3O2S2 | 10 MG
SDP
Catalog No. 5000286978
Encompass_Preferred
CDK1/Cyc B-IN-1 is a small-molecule inhibitor selective for the CDK1-cyclin B kinase complex. It inhibits CDK1-cyclin B activity (reported IC50 ≈97 nM) and induces G2/M cell-cycle arrest and apoptosis in cancer cell lines. The compound is supplied as a solid with high purity and is used in biochemical and cellular studies of cell-cycle regulation.
  • Selective CDK1-cyclin B inhibitor with reported IC50 ≈97 nM.
  • Induces G2/M cell-cycle arrest and apoptosis in cell lines.
  • Suitable for biochemical kinase assays and cell-based studies.
  • High supplied purity for reproducible results.
  • Soluble in DMSO at 100 mg/mL; use newly opened DMSO.
  • Stable as a powder under recommended cold storage conditions.
Catalog No. 50-002-86978 Supplier Medchemexpress LLC Supplier No. HY14764610MG
CDK1/Cyc B-IN-1 is a small-molecule inhibitor selective for the CDK1-cyclin B kinase complex. It inhibits CDK1-cyclin B activity (reported IC50 ≈97 nM) and induces G2/M cell-cycle arrest and apoptosis in cancer cell lines. The compound is supplied as a solid with high purity and is used in biochemical and cellular studies of cell-cycle regulation.
  • Selective CDK1-cyclin B inhibitor with reported IC50 ≈97 nM.
  • Induces G2/M cell-cycle arrest and apoptosis in cell lines.
  • Suitable for biochemical kinase assays and cell-based studies.
  • High supplied purity for reproducible results.
  • Soluble in DMSO at 100 mg/mL; use newly opened DMSO.
  • Stable as a powder under recommended cold storage conditions.