Learn More
Medchemexpress LLC 7-chloro-3-methyl-3,4-dihydro-2H-1,2,4-benzothiadiazine 1,1-dioxide | 22503-72-6 | 99.4% | 232.69 g/mol | C8H9ClN2O2S | 25 MG
SDP
Catalog No. 5000246198
Encompass_Preferred
IDRA 21 is a benzothiadiazine derivative that functions as a positive allosteric modulator of AMPA receptors. It enhances excitatory neurotransmission via GluR1/2-containing AMPA receptors and is used as a research reagent in studies of cognition, memory, and synaptic plasticity.
  • Enhances AMPA receptor-mediated excitatory neurotransmission.
  • Useful for cognition and memory research in preclinical models.
  • Suitable for biochemical and pharmacological assays at research scale.
  • High reported purity supports reproducible experimental results.
Catalog No. 50-002-46198 Supplier Medchemexpress LLC Supplier No. HY10152825MG
IDRA 21 is a benzothiadiazine derivative that functions as a positive allosteric modulator of AMPA receptors. It enhances excitatory neurotransmission via GluR1/2-containing AMPA receptors and is used as a research reagent in studies of cognition, memory, and synaptic plasticity.
  • Enhances AMPA receptor-mediated excitatory neurotransmission.
  • Useful for cognition and memory research in preclinical models.
  • Suitable for biochemical and pharmacological assays at research scale.
  • High reported purity supports reproducible experimental results.