Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-chloro-3-methyl-3,4-dihydro-2H-1,2,4-benzothiadiazine 1,1-dioxide | 22503-72-6 | MFCD00270874 | 99.4% | 232.69 g/mol | C8H9ClN2O2S | 1 ML
SDP

Catalog No. 5000248797
Encompass_Preferred

IDRA-21 is a benzothiadiazine derivative that acts as a positive allosteric modulator of AMPA receptors. It inhibits AMPA receptor desensitization and is used in neuroscience research related to cognition, memory, and excitatory neurotransmission. The compound is supplied as a 10 mM solution in DMSO (1 mL) and is also available as solid quantities; reported purity is approximately 99.4%.

  • Positive allosteric modulator of AMPA receptors.
  • Inhibits AMPA receptor desensitization to enhance excitatory signaling.
  • Useful for research on cognition and memory mechanisms.
  • Available as a 10 mM solution in DMSO (1 mL) and in multiple solid sizes.
  • High reported purity (~99.4%).
  • Molecular weight 232.69 g/mol; formula C8H9ClN2O2S.

Catalog No. 50-002-48797 Supplier Medchemexpress LLC Supplier No. HY10152810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IDRA-21 is a benzothiadiazine derivative that acts as a positive allosteric modulator of AMPA receptors. It inhibits AMPA receptor desensitization and is used in neuroscience research related to cognition, memory, and excitatory neurotransmission. The compound is supplied as a 10 mM solution in DMSO (1 mL) and is also available as solid quantities; reported purity is approximately 99.4%.

  • Positive allosteric modulator of AMPA receptors.
  • Inhibits AMPA receptor desensitization to enhance excitatory signaling.
  • Useful for research on cognition and memory mechanisms.
  • Available as a 10 mM solution in DMSO (1 mL) and in multiple solid sizes.
  • High reported purity (~99.4%).
  • Molecular weight 232.69 g/mol; formula C8H9ClN2O2S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.