Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Chloro-4-hydroxyquinoline 97.0+%, TCI America™
SDP

Catalog No. C109210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C109210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C109210G Supplier TCI America Supplier No. C109210G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 86-99-7
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00006778
InChI Key XMFXTXKSWIDMER-UHFFFAOYSA-N
Synonym 7-chloroquinolin-4-ol, 7-chloro-4-hydroxyquinoline, 7-chloro-4-quinolinol, 7-chloroquinolin-4 1h-one, 4-quinolinol, 7-chloro, 4 1h-quinolinone, 7-chloro, 7-chloro-quinolin-4-ol, 7-chloro-4 1h-quinolinone, 4mrw, 7-chloro4-quinolinol
PubChem CID 66593
IUPAC Name 7-chloro-1H-quinolin-4-one
SMILES C1=CC2=C(C=C1Cl)NC=CC2=O

Specifications

Melting Point 279°C
Color Yellow
Quantity 10 g
Formula Weight 179.60
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Chloro-4-hydroxyquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.