Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-chloro-4-(piperazin-1-yl)quinoline | 837-52-5 | MFCD01476123 | 98.6% | 247.72 g/mol | C13H14ClN3 | 100 MG
SDP

Catalog No. 5000300468
Encompass_Preferred

7-Chloro-4-(piperazin-1-yl)quinoline is a small-molecule quinoline scaffold bearing a piperazine substituent, supplied as a solid for chemical research and screening applications. It is characterized by a defined molecular formula and molecular weight, and is provided with quality documentation for laboratory use.

  • Small-molecule quinoline scaffold suitable for medicinal chemistry and screening.
  • Molecular formula C13H14ClN3 and molecular weight 247.72 g/mol.
  • White to yellow solid with high purity (98.6%).
  • Supplied in small pack sizes appropriate for research workflows.
  • Storage stability: powder stable at -20°C for long-term storage.
  • Documentation available: COA, SDS, and handling instructions.

Catalog No. 50-003-00468 Supplier Medchemexpress LLC Supplier No. HYW020111100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Chloro-4-(piperazin-1-yl)quinoline is a small-molecule quinoline scaffold bearing a piperazine substituent, supplied as a solid for chemical research and screening applications. It is characterized by a defined molecular formula and molecular weight, and is provided with quality documentation for laboratory use.

  • Small-molecule quinoline scaffold suitable for medicinal chemistry and screening.
  • Molecular formula C13H14ClN3 and molecular weight 247.72 g/mol.
  • White to yellow solid with high purity (98.6%).
  • Supplied in small pack sizes appropriate for research workflows.
  • Storage stability: powder stable at -20°C for long-term storage.
  • Documentation available: COA, SDS, and handling instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.