Learn More
Medchemexpress LLC 7-chloro-4-(piperazin-1-yl)quinoline | 837-52-5 | MFCD01476123 | 98.6% | 247.72 g/mol | C13H14ClN3 | 25 MG
SDP
Catalog No. 5000303801
Encompass_Preferred
7-chloro-4-(piperazin-1-yl)quinoline is a small-molecule quinoline scaffold used in medicinal chemistry and drug discovery as a building block and intermediate. It contains a piperazinyl substituent at the 4-position and a chloro substituent at the 7-position, providing a versatile core for SAR exploration and synthetic derivatization. Supplied as a solid with reported high purity, it is intended for research use only.
  • High purity for reliable synthetic and analytical results.
  • Quinoline scaffold amenable to functionalization at multiple positions.
  • Piperazinyl substituent supports structure-activity relationship work.
  • Solid form enables straightforward weighing and storage.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-003-03801 Supplier Medchemexpress LLC Supplier No. 25MGHYW020111
7-chloro-4-(piperazin-1-yl)quinoline is a small-molecule quinoline scaffold used in medicinal chemistry and drug discovery as a building block and intermediate. It contains a piperazinyl substituent at the 4-position and a chloro substituent at the 7-position, providing a versatile core for SAR exploration and synthetic derivatization. Supplied as a solid with reported high purity, it is intended for research use only.
  • High purity for reliable synthetic and analytical results.
  • Quinoline scaffold amenable to functionalization at multiple positions.
  • Piperazinyl substituent supports structure-activity relationship work.
  • Solid form enables straightforward weighing and storage.
  • Intended for research use only; not for human or veterinary use.