Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-[(Chlorocarbonyl)methoxy]-4-methylcoumarin 98%, Thermo Scientific™

Catalog No. AC404585000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC404585000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC404585000 Supplier Thermo Scientific Chemicals Supplier No. 404585000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91454-65-8
Molecular Formula C12H9ClO4
Molecular Weight (g/mol) 252.65
InChI Key CTSWCAAFHPVFFO-UHFFFAOYSA-N
Synonym 7-chlorocarbonylmethoxy-4-methylcoumarin, 7-chlorocarbonyl methoxy-4-methylcoumarin, 7-ccmmc, 2-4-methyl-2-oxochromen-7-yloxy acetyl chloride, 2-4-methyl-2-oxochromen-7-yl oxy acetyl chloride, 7-ccm-4-mc, acetyl chloride, 4-methyl-2-oxo-2h-1-benzopyran-7-yl oxy-, s, 7-chlorocar-bonyl methoxy-4-methylcoumarin, 2-4-methyl-2-oxochromen-7-yl oxyacetyl chloride, 4-methyl-2-oxobenzo b pyran-7-yloxy acetyl chloride
PubChem CID 124645
IUPAC Name 2-(4-methyl-2-oxochromen-7-yl)oxyacetyl chloride
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OCC(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Blue-Gray to Blue-Green
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 500 mg
Formula Weight 252.65
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 7-[(Chlorocarbonyl)methoxy]-4-methylcoumarin, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.