Learn More
Medchemexpress LLC 7-chlorokynurenic acid | 18000-24-3 | MFCD00069227 | 223.61 | C10H6ClNO3 | 5 MG
SDP
Catalog No. 5000212969
Encompass_Preferred
7-Chlorokynurenic acid is a research tool compound used to probe NMDA receptor function by selectively antagonizing the glycine coagonist site. It also inhibits vesicular glutamate uptake and has reported antinociceptive effects in certain delivery models, making it useful for neuropharmacology and synaptic signaling studies.
  • Potent and selective antagonist of NMDA receptor glycine site.
  • Inhibits glutamate reuptake into synaptic vesicles.
  • Suitable for in vitro and in vivo research applications.
  • Supplied as a powder for flexible preparation and dosing.
  • Stable under frozen storage for extended periods when handled per datasheet.
Catalog No. 50-002-12969 Supplier Medchemexpress LLC Supplier No. HY1008115MG
7-Chlorokynurenic acid is a research tool compound used to probe NMDA receptor function by selectively antagonizing the glycine coagonist site. It also inhibits vesicular glutamate uptake and has reported antinociceptive effects in certain delivery models, making it useful for neuropharmacology and synaptic signaling studies.
  • Potent and selective antagonist of NMDA receptor glycine site.
  • Inhibits glutamate reuptake into synaptic vesicles.
  • Suitable for in vitro and in vivo research applications.
  • Supplied as a powder for flexible preparation and dosing.
  • Stable under frozen storage for extended periods when handled per datasheet.