Learn More
Medchemexpress LLC 7-chlorokynurenic acid | 18000-24-3 | MFCD00069227 | 98.0% | 223.61 g·mol-1 | C10H6ClNO3 | 100 MG
SDP
Catalog No. 5000212967
Encompass_Preferred
7-Chlorokynurenic acid is a research-grade small molecule that potently and selectively antagonizes the glycine co-agonist site of the NMDA receptor. Supplied as a powder, it is used in neuropharmacology and electrophysiology studies to probe NMDA receptor function.
  • Potent and selective antagonist of the NMDA receptor glycine site.
  • Reported IC50 approximately 0.56 μM for glycine-site inhibition.
  • Powder form suitable for preparation of DMSO stock solutions.
  • Good solubility in DMSO (≈14.29 mg/mL) and insoluble in water.
  • Well characterized chemical identity (CAS 18000-24-3; MW 223.61 g·mol-1).
Catalog No. 50-002-12967 Supplier Medchemexpress LLC Supplier No. HY100811100MG
7-Chlorokynurenic acid is a research-grade small molecule that potently and selectively antagonizes the glycine co-agonist site of the NMDA receptor. Supplied as a powder, it is used in neuropharmacology and electrophysiology studies to probe NMDA receptor function.
  • Potent and selective antagonist of the NMDA receptor glycine site.
  • Reported IC50 approximately 0.56 μM for glycine-site inhibition.
  • Powder form suitable for preparation of DMSO stock solutions.
  • Good solubility in DMSO (≈14.29 mg/mL) and insoluble in water.
  • Well characterized chemical identity (CAS 18000-24-3; MW 223.61 g·mol-1).