Learn More
Medchemexpress LLC 7-chlorokynurenic acid | 18000-24-3 | MFCD00069227 | 99.9% | 223.61 g/mol | C10H6ClNO3 | 1 ML
SDP
Catalog No. 5000212968
Encompass_Preferred
7-Chlorokynurenic acid is a selective antagonist of the glycine co-agonist site of the NMDA receptor. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for research use in neuropharmacology and receptor biology; handle and store according to safety guidance.
Catalog No. 50-002-12968 Supplier Medchemexpress LLC Supplier No. HY10081110MM 1ML
7-Chlorokynurenic acid is a selective antagonist of the glycine co-agonist site of the NMDA receptor. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for research use in neuropharmacology and receptor biology; handle and store according to safety guidance.