Learn More
Cayman Chemical 7-Chlorokynurenic Acid, Purity: ≥95%, CAS: 18000-24-3, 50 mg
SDP
Catalog No. 502948980
Encompass_Preferred
A glycine-site selective NMDA receptor antagonist; selectively binds to the NMDA receptor glycine-binding site (IC50 = 0.56 µM) over the NMDA-, quisqualate-, and kainate-binding sites (IC50s = 169, 153, and >1,000 µM, respectively); induces antidepressant-like activity in a variety of behavioral tests in mice at 0.1 and 1 mg/kg
Catalog No. 50-294-8980 Supplier Cayman Chemical Supplier No. 3340850 MG
A glycine-site selective NMDA receptor antagonist; selectively binds to the NMDA receptor glycine-binding site (IC50 = 0.56 µM) over the NMDA-, quisqualate-, and kainate-binding sites (IC50s = 169, 153, and >1,000 µM, respectively); induces antidepressant-like activity in a variety of behavioral tests in mice at 0.1 and 1 mg/kg