Learn More
Medchemexpress LLC HY-100811A 10mg Medchemexpress, 7-Chlorokynurenic acid sodium salt CAS:1263094-00-3 Purity:>98%
SDP
Catalog No. 501931960
Encompass_Preferred
Medchemexpress, HY-100811A 10mg 7-Chlorokynurenic acid sodium salt CAS:1263094-00-3 7-Chlorokynurenic acid sodium salt is a potent and selective antagonist of the glycine B coagonist site of the N-methyl-D-aspartate (NMDA) receptor ( IC 50 =0.56 μM). 7-Chlorokynurenic acid sodium salt is also a potent inhibitor of the reuptake of glutamate into synaptic vesicles with a K i of 0.59 μM. 7-Chlorokynurenic acid sodium salt has potent antinociceptive actions after neuraxial delivery [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1960 Supplier Medchemexpress LLC Supplier No. HY100811A10MG
Medchemexpress, HY-100811A 10mg 7-Chlorokynurenic acid sodium salt CAS:1263094-00-3 7-Chlorokynurenic acid sodium salt is a potent and selective antagonist of the glycine B coagonist site of the N-methyl-D-aspartate (NMDA) receptor ( IC 50 =0.56 μM). 7-Chlorokynurenic acid sodium salt is also a potent inhibitor of the reuptake of glutamate into synaptic vesicles with a K i of 0.59 μM. 7-Chlorokynurenic acid sodium salt has potent antinociceptive actions after neuraxial delivery [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.