Learn More
Abcam 7-Chlorokynurenic acid sodium salt, NMDA receptor glycine site antagonist, 10MG
Catalog No. 502584642
Encompass_Preferred
MW 245.59 Da, Purity >99%. Potent NMDA receptor glycine site antagonist. Water soluble form. Also available in simple stock solutions (ab146692) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4642 Supplier Abcam Supplier No. AB12025510MG
Add to Cart
Add to Cart
MW 245.59 Da, Purity >99%. Potent NMDA receptor glycine site antagonist. Water soluble form. Also available in simple stock solutions (ab146692) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement